missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Multi-Element Coolant Standard: P @ 2500; B, K, Na @ 1000; Mo, Si @ 500; Al, Ca, Cu, Fe, Mg, Pb, Zn @ 50 μg/g in dil. HNO3, tr. HF, VHG™
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
Description
Multi-Element Coolant Standard: P @ 2500; B, K, Na @ 1000; Mo, Si @ 500; Al, Ca, Cu, Fe, Mg, Pb, Zn @ 50 μg/g in dil. HNO3, tr. HF
Specifications
Specifications
| Chemical Name or Material | Aluminium/Calcium/Copper/Iron/Lead/Magnesium/Zinc/Molybdenum/Silicon/Boron/Potassium/Sodium/Phosphorus |
| CAS | 7429-90-5, 7440-70-2, 7440-50-8, 7439-89-6, 7439-92-1, 7439-95-4 |
| Methods | ICP |
| Analyte or Component Names | Aluminium/Calcium/Copper/Iron/Lead/Magnesium/Zinc/Molybdenum/Silicon/Boron/Potassium/Sodium/Phosphorus |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Aluminium 50 μg/g/Calcium 50 μg/g/Copper 50 μg/g/Iron 50 μg/g/Lead 50 μg/g/Magnesium 50 μg/g/Zinc 50 μg/g/Molybdenum 500 μg/g/Silicon 500 μg/g/Boron 1000 μg/g/Potassium 1000 μg/g/Sodium 1000 μg/g/Phosphorus 2500 μg/g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction