missing translation for 'onlineSavingsMsg'
Learn More

Multi-Element Calibration Standard 3 for ICP, Fluka™

Product Code. 16646222 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16646222 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16646222 Supplier Honeywell-Fluka Supplier No. E2027100ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Certified Reference Material, 10 mg/L each of Sb, Au, Hf, Ir, Pd, Pt, Rh, Ru, Te, Sn in HCl 10% / HNO3 1%

Honeywell Fluka™ analytical standards offers a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. Our products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Multi-Element Calibration Standard 3 for ICP
Packaging Plastic Bottle
CAS 16950-06-4
Technique ICP-OES and ICP-MS
pH <2
Quantity 100 mL
Grade ICP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.