Learn More
BD Biosciences™ Multi-Check Control (CE/IVD)
Product Code. 15855938
Click to view available options
Quantity:
1 x 2.5 mL
2 x 2.5 mL
5 x 2.5 mL
Unit Size:
Each
Product Code. 15855938
Brand: BD Biosciences™ 340912
Intended as a complete process control for immunophenotyping by flow cytometry.
  • Complete process control for immunophenotyping by flow cytometry.
  • Control for antibody staining, red blood cell (RBC) lysis, instrument setup and performance, and data analysis.
  • Stable control with assigned values that can be used to monitor the immunophenotyping process.
  • The BD Multi-Check Control should be treated in the same manner as whole blood.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance IVD 2/2014, CE 23-5203-04
Content And Storage Store vials upright, tightly capped, at 2°C to 8°C.
Quantity 2 x 2.5 mL
Stability Unopened vials are stable until the expiration date. Opened vials are stable for 9 thermal cycles when handled properly.