missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ Multi-Check CD4 Low Control (CE/IVD)
Click to view available options
Quantity:
1 x 2.5 mL
2 x 2.5 mL
5 x 2.5 mL
Unit Size:
Each
Description
- Complete process control for immunophenotyping by flow cytometry.
- Control for antibody staining, red blood cell (RBC) lysis, instrument setup and performance, and data analysis.
- Stable control with assigned values that can be used to monitor the immunophenotyping process.
- The BD Multi-Check CD4 Low Control should be treated in the same manner as whole blood.
Specifications
Specifications
| Certifications/Compliance | IVD 2/2014, CE 23-5208-04 |
| Content And Storage | Store vials upright, tightly capped, at 2°C to 8°C. |
| Quantity | 5 x 2.5 mL |
| Stability | Unopened vials are stable until the expiration date. Opened vials are stable for 9 thermal cycles when handled properly. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction