missing translation for 'onlineSavingsMsg'
Learn More

Multi-Cation Standard 1: Li+ @ 50; Na+ @ 200; Mg+2, NH4+ @ 250; Ca+2, K+ @ 500 μg/mL in dil. HNO3, VHG™

Product Code. 30003125
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003125 500 mL 500mL
30003151 250 mL 250mL
30003159 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003125 Supplier VHG Labs Supplier No. VHGICM5A500
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Multi-Cation Standard 1: Li+ @ 50; Na+ @ 200; Mg+2, NH4+ @ 250; Ca+2, K+ @ 500 μg/mL in dil. HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium/Sodium/Ammonium/Magnesium/Calcium/Potassium
CAS 7439-93-2, 7440-23-5, 14798-03-9, 7439-95-4, 7440-70-2, 9-7-7440
Methods Ion Chromatography
Analyte or Component Names Lithium/Sodium/Ammonium/Magnesium/Calcium/Potassium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Lithium 50 μg/mL /Sodium 200 μg/mL /Ammonium 250 μg/mL /Magnesium 250 μg/mL /Calcium 500 μg/mL /Potassium 500 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.