missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QIAGEN Multi-Barrier Pouches
Click to view available options
Size:
Medium (11.1 x 16.5 cm)
Small (9.5 x 7.6 cm)
Unit Size:
Pack of 100
Description
For contamination-free collection, storage and transport of DNA samples.
- Multi-barrier Pouches for transport or storage of QIAcard FTA Mini and Micro formats.
- Laminated layers maintain sample integrity and security by protecting the card from exposure to gas or liquid contamination.
- Maintain sample security by tamper-evident seal.
- The outer paper surface is suitable for labeling or writing.
- To optimize sample integrity FTA Cards are stored in a Multi-barrier Pouch with a desiccant packet.
Specifications
Specifications
| For Use With (Equipment) | FTA™ gene card, mini card or micro card |
| For Use With (Application) | Collection, storage, processing and shipping FTA™ and FTA™ elute cards |
| Product Type | Multi-barrier Pouch |
| Quantity | 100 Pk. |
| Size | Small (9.5 x 7.6 cm) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction