missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Multi-Anion Standard 3: F- @ 20; Cl- @ 30; NO3- @ 100; PO4(-3), SO4(-2) @ 150 μg/mL in H2O, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
Multi-Anion Standard 3: F- @ 20; Cl- @ 30; NO3- @ 100; PO4(-3), SO4(-2) @ 150 μg/mL in H2O
Specifications
Specifications
| Chemical Name or Material | Fluoride/Chloride/Nitrate/Phosphate/Sulfate |
| CAS | 16984-48-8, 16887-00-6, 14797-55-8, 14265-44-2, 14808-79-8 |
| Methods | Ion Chromatography |
| Analyte or Component Names | Fluoride/Chloride/Nitrate/Phosphate/Sulfate |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Water |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Fluoride 20 μg/mL/Chloride 30 μg/mL/Nitrate 100 μg/mL/Phosphate 150 μg/mL/Sulfate 150 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction