missing translation for 'onlineSavingsMsg'
Learn More

Multi 13 elements solution - Be,Na,Mg,K,Ca,Fe,Co,Rh,In,Ba,Ce,Pb,U for ICP 0.001mg/L, Fisher chemical™

Product Code. p-7083775 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
This item is not returnable. View return policy

Product Code. 12956183

Brand: Fisher Chemical MS 003 250

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Chemical Name or Material Thirteen Elements
Packaging Bottle
Analyte or Component Names Be,Na,Mg,K,Ca,Fe,Co,Rh,In,Ba,Ce,Pb,U
Physical Form Liquid
Concentration 0.001 mg/L
Type Limit Detection Multi-component Verification Solution
Element Be, Na, Mg, K, Ca, Fe, Co, Rh, In, Ba, Ce, Pb, U
Solvent or Matrix 2% HNO3
Quantity 250 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.