missing translation for 'onlineSavingsMsg'
Learn More

Mullite

Product Code. 11378107
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378107 500 g 500g
11388107 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378107 Supplier Thermo Scientific Alfa Aesar Supplier No. 022881.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1302-93-8 | Al6O13Si2 | 426.05 g/mol

Mullite is present in the form of needles in porcelain, it is used in high-mullite based products is in hot blast stove checker bricks, melting and firing processes, and is used as a refractory material, manufacture of a type of crucible, steel industry, electronic substrates and protective coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1302-93-8
Molecular Formula Al6O13Si2
Molecular Weight (g/mol) 426.05
MDL Number MFCD00058866
InChI Key KZHJGOXRZJKJNY-UHFFFAOYSA-N
Synonym Aluminum oxide/silicon oxide

Specifications

Color White
Physical Form Powder
Quantity 500 g
Solubility Information Insoluble in water.
Odor Odorless
Chemical Name or Material Mullite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.