missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sensititre™ Cation Adjusted Mueller-Hinton Broth w/ TES w/ Lysed Horse Blood

Product Code. 15405504
Change view
Click to view available options
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
15405504 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15405504 Supplier Thermo Scientific™ Supplier No. YCP11210
  • ONLINE EXCLUSIVE PRICE
    £77.30
    £70.36 / Pack of 10
    Save £6.94
    8% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform susceptibility testing with a full range of supplies, including this broth, to complement and enhance your Thermo Scientific™ Sensititre™ test program.

Perform high-quality susceptibility testing with a complete range of supplies, including Thermo Scientific™ Sensititre™ Cation Adjusted Mueller-Hinton Broth w/ TES w/ Lysed Horse Blood, to complement and enhance your Thermo Scientific™ Sensititre™ test program.

Not all products are available for sale in all territories. Please inquire. 

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Format Tube
Manufacturing Origin United Kingdom
Media Type Mueller Hinton LHB Cation Adjusted
Product Type Microbiology Media
Test Type Manual Diagnostics
For Use With (Application) Trek Sensititre
No. per Pack 10
Packaging Type 10 x 11 mL Tubes
Species Multiple
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.