missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mucolipin 1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-82375UV
This item is not returnable.
View return policy
Description
Mucolipin 1 Polyclonal specifically detects Mucolipin 1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Mucolipin 1 | |
| Polyclonal | |
| Western Blot | |
| MG-2, ML4TRP-ML1, MLIV, MST080, MSTP080, mucolipidin, mucolipidosis type IV protein, mucolipin 1, mucolipin-1, TRPML1, TRP-ML1, TRPM-L1 | |
| Rabbit | |
| Affinity Purified | |
| Cell Biology, Neuroscience, Sensory Systems, Signal Transduction, Vision | |
| 57192.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| DyLight 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MCOLN1 | |
| Synthetic peptide made to a C-terminal portion of mouse Mucolipin 1 (within residues 500-580). [Swiss-ProtNo. Q99J21] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction