missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mucochloric Acid, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | Mucochloric Acid |
| CAS | 87-56-9 |
| Molecular Formula | C4H2Cl2O3 |
| Quantity | 50 g |
| Synonym | (Z)-2,3-Dichloro-4-oxo-2-butenoic Acid, Dichloromalealdehydic Acid, 2,3-Dichloromaleic Aldehyde Acid, Dichloromalealdehydic Acid, α,β-Dichloro-β-formylacrylic Acid |
| InChI Formula | InChI=1S/C4H2Cl2O3/c5-2(1-7)3(6)4(8)9/h1H,(H,8,9)/b3-2- |
| SMILES | OC(=O)\C(=C(\Cl)/C=O)\Cl |
| IUPAC Name | (Z)-2,3-dichloro-4-oxobut-2-enoic acid |
| Molecular Weight (g/mol) | 168.96 |
| Formula Weight | 167.9381 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction