missing translation for 'onlineSavingsMsg'
Learn More

Mucochloric acid, 99% (dry wt.), water <4.0%, Thermo Scientific Chemicals

Product Code. 11490348
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490348 500 g 500g
11480348 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490348 Supplier Thermo Scientific Alfa Aesar Supplier No. B24523.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-56-9
Molecular Formula C4H2Cl2O3
Molecular Weight (g/mol) 168.96
MDL Number MFCD00006966,MFCD00006966
InChI Key LUMLZKVIXLWTCI-IHWYPQMZSA-N
Synonym mucochloric acid, 2,3-dichloromaleic aldehyde acid, kyselina mukochlorova, 2z-2,3-dichloro-4-oxobut-2-enoic acid, 2-butenoic acid, 2,3-dichloro-4-oxo-, 2z, aldehydodichloromaleic acid, dichloromalealdehydic acid, unii-5i5877jhiw, kyselina mukochlorova czech, ccris 6597
PubChem CID 2771871
SMILES OC(=O)C(\Cl)=C(\Cl)C=O

Specifications

Melting Point 125°C to 128°C (dried)
Density 1.85
Flash Point 186°C (367°F)
Quantity 500 g
UN Number UN2923
Beilstein 1705641
Merck Index 14,6297
IUPAC Name (Z)-2,3-dichloro-4-oxobut-2-enoic acid
Formula Weight 168.96
Percent Purity 99%
Assay (dry wt.), water <4.0%
Chemical Name or Material Mucochloric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.