missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mucin 4X/Mucin 4Y Recombinant Rabbit Monoclonal Antibody (HL2026)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA550363

Product Code. 18015631

  • £405.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Mucin 4X/Mucin 4Y Recombinant Monoclonal Antibody for Western Blot, ICC/IF

Mucin 4X/Mucin 4Y (MUC4) is a gene target that codes for a transmembrane glycoprotein expressed in various epithelial tissues. MUC4 plays an important role in cellular adhesion, signaling, and protection of the epithelial surface. It is often overexpressed in various cancers, including pancreatic, ovarian, and breast cancers, and has been identified as a potential biomarker for these cancers. Targeting MUC4 has shown promise as a therapeutic strategy for cancer treatment.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mucin 4X/Mucin 4Y
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Q99102
MUC4
Recombinant fragment of human Mucin 4 X+Mucin 4 Y.
100 μL
Primary
Human
Antibody
IgG
Western Blot, Immunocytochemistry
HL2026
Unconjugated
MUC4
Mucin-4X/Mucin-4Y
Rabbit
Protein A
RUO
4585
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.