missing translation for 'onlineSavingsMsg'
Learn More

MUCDHL Antibody [DyLight 550], Novus Biologicals™

Product Code. 30655297 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30655297 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30655297 Supplier Novus Biologicals Supplier No. AF4159L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

MUCDHL Polyclonal antibody specifically detects MUCDHL in Mouse samples. It is validated for Flow Cytometry,Western Blot,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUCDHL
Applications Flow Cytometry, Western Blot, CyTOF
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias cadherin-related family member 5, FLJ20219, MUCDHLmucin-like protocadherin, mucin and cadherin-like, Mucin and cadherin-like protein, MUPCDH, MU-PCDH, mu-protocadherin
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse MUCDHL, Gln29-Ala479, Accession # NP_082345
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53841
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.