missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MUC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103932
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MUC7.
This gene encodes a small salivary mucin, which is thought to play a role in facilitating the clearance of bacteria in the oral cavity and to aid in mastication, speech, and swallowing. The central domain of this glycoprotein contains tandem repeats, each composed of 23 amino acids. This antimicrobial protein has antibacterial and antifungal activity. The most common allele contains 6 repeats, and some alleles may be associated with susceptibility to asthma. Alternatively spliced transcript variants with different 5' UTR, but encoding the same protein, have been found for this gene.
Specifications
| MUC7 | |
| Polyclonal | |
| Unconjugated | |
| MUC7 | |
| apo-MG2; MG2; MUC7; MUC-7; mucin 7, salivary; mucin 7, secreted; mucin-7; Salivary mucin-7 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4589 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8TAX7 | |
| MUC7 | |
| A synthesized peptide derived from human MUC7(Accession Q8TAX7), corresponding to amino acid residues S62-P112. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction