missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC7 Antibody (4D2-1D7), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50391H
This item is not returnable.
View return policy
Description
MUC7 Monoclonal specifically detects MUC7 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| MUC7 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| MUC7 | |
| KLH conjugated peptide EGRERDHELRHRRHHHQ located in the N-terminal part of the MUC7 protein within the histatin-like domain.was used to immunise Balb/c female mice. | |
| 0.1 mL | |
| Primary | |
| No cross reactivity with other human mucins. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| 4D2-1D7 | |
| Western Blot, ELISA, Immunohistochemistry | |
| apo-MG2, DKFZp686J03256, FLJ27047, MG2MGC34772, MUC-7, mucin 7, secreted, mucin-7, salivary, Salivary mucin-7 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 4589 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction