missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC6 Rabbit anti-Human, Clone: MUC6/1553R, Novus Biologicals™
Rabbit Monoclonal Antibody
£245.00
Specifications
| Antigen | MUC6 |
|---|---|
| Clone | MUC6/1553R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 2-4 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
Description
MUC6 Monoclonal specifically detects MUC6 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| MUC6 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| gastric mucin, Gastric mucin-6, MUC-6, mucin 6, gastric, mucin 6, oligomeric mucus/gel-forming, mucin-6 | |
| Recombinant full-length human MUC6 protein | |
| Primary | |
| Store at 4°C. |
| MUC6/1553R | |
| Immunohistochemistry-Paraffin 2-4 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 4588 | |
| IgG | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title