missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC5B Antibody (6F10-E4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£169.00 - £329.00
Specifications
| Antigen | MUC5B |
|---|---|
| Clone | 6F10-E4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18690067
|
Novus Biologicals
NBP2-50390-0.025ml |
0.025 mL |
£169.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623269
|
Novus Biologicals
NBP2-50390 |
0.1 mL |
£329.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MUC5B Monoclonal antibody specifically detects MUC5B in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| MUC5B | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, Cellular Signaling | |
| PBS | |
| 727897 | |
| IgG1 κ | |
| Protein A purified |
| 6F10-E4 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| high molecular weight salivary mucin MG1, mucin 5, subtype B, tracheobronchial, mucin 5B, oligomeric mucus/gel-forming, sublingual gland mucin, tracheobronchial | |
| KLH conjugated peptide RNREQVGKF in 4 of the cysteine domains of the tandem repeat region was used to immunise Balb/c female mice. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title