missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC5B Antibody (6F10-E4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50390B
This item is not returnable.
View return policy
Description
MUC5B Monoclonal specifically detects MUC5B in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Mucin 5B | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| MUC5B | |
| KLH conjugated peptide RNREQVGKF in 4 of the cysteine domains of the tandem repeat region was used to immunise Balb/c female mice. | |
| 0.1 mL | |
| Cell Cycle and Replication, Cellular Signaling | |
| 727897 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| 6F10-E4 | |
| Western Blot, ELISA, Immunohistochemistry | |
| high molecular weight salivary mucin MG1, mucin 5, subtype B, tracheobronchial, mucin 5B, oligomeric mucus/gel-forming, sublingual gland mucin, tracheobronchial | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction