missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC5B Antibody (19.4E), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-29706CL1
This item is not returnable.
View return policy
Description
MUC5B Monoclonal antibody specifically detects MUC5B in Human,Mouse (Negative),Primate samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| MUC5B | |
| Monoclonal | |
| CoraFluor 1 | |
| high molecular weight salivary mucin MG1, mucin 5, subtype B, tracheobronchial, mucin 5B, oligomeric mucus/gel-forming, sublingual gland mucin, tracheobronchial | |
| Un-glycosylated, serine- and threonine-free synthetic peptide from the N-terminus of human MUC5B. | |
| 0.1 mL | |
| Cell Cycle and Replication, Cellular Signaling | |
| 727897 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 19.4E | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse (Negative), Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction