missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC5AC Antibody (MUC5AC/917 + 45M1), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47696B
This item is not returnable.
View return policy
Description
MUC5AC Monoclonal specifically detects MUC5AC in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| Mucin 5AC | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| MUC5AC | |
| Recombinant human MUC5AC protein (MUC5AC/917); M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient (45M1) (Uniprot: P98088) | |
| 0.1 mL | |
| Primary | |
| Mucin 5AC glycoprotein (MUC5AC) is a 641kDa glycoprotein belonging to the superfamily of mucins. Mucins are high molecular weight glycoproteins produced by epithelial cells and can be divided into two families; secretory mucins and membrane bound mucins. MUC5AC is a mucus-forming secreted mucin that is found in normal gastric and tracheo-bronchial mucosa, but absent from normal colon. MUC5AC expression is present in primary ovarian mucinous cancer but usually absent in colorectal adenocarcinoma, thus showing an expression pattern opposite to MUC2. Together with a panel of antibodies, Anti-MUC5AC may be useful for differential identification of primary mucinous ovarian tumors from colon adenocarcinoma metastatic to the ovary. MUC5AC antibodies may also be useful for identification of intestinal metaplasia as well as in the identification of pancreatic carcinoma and pre-cancerous changes vs. normal pancreas. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| MUC5AC/917 + 45M1 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence | |
| gastric mucin, leB, lewis B blood group antigen, major airway glycoprotein, MUC5, mucin 5, subtypes A and C, tracheobronchial/gastric, mucin 5AC, oligomeric mucus/gel-forming, mucin 5AC, oligomeric mucus/gel-forming pseudogene, mucin-5 subtype AC, tracheobronchial, mucin-5AC, TBM, tracheobronchial mucin | |
| Mouse | |
| Protein A or G purified | |
| Cellular Markers, Extracellular Matrix, Infections (Virus Bacteria and Parasites), Signal Transduction | |
| 4586 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction