missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MUC5AC Monoclonal antibody specifically detects MUC5AC in Human, Mouse, Rat, Bovine, Chicken, Feline, Monkey, Rabbit samples. It is validated for ELISA
Specifications
Specifications
| Antigen | MUC5AC |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1-13M1 |
| Conjugate | Alexa Fluor 700 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | gastric mucin, leB, lewis B blood group antigen, major airway glycoprotein, MUC5, mucin 5, subtypes A and C, tracheobronchial/gastric, mucin 5AC, oligomeric mucus/gel-forming, mucin 5AC, oligomeric mucus/gel-forming pseudogene, mucin-5 subtype AC, tracheobronchial, mucin-5AC, TBM, tracheobronchial mucin |
| Host Species | Mouse |
| Immunogen | M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?