missing translation for 'onlineSavingsMsg'
Learn More

MUC5AC Antibody (1-13M1), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18324614 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18324614 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18324614 Supplier Novus Biologicals Supplier No. NBP300536AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MUC5AC Monoclonal antibody specifically detects MUC5AC in Human, Mouse, Rat, Bovine, Chicken, Feline, Monkey, Rabbit samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUC5AC
Applications ELISA
Classification Monoclonal
Clone 1-13M1
Conjugate Alexa Fluor 594
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias gastric mucin, leB, lewis B blood group antigen, major airway glycoprotein, MUC5, mucin 5, subtypes A and C, tracheobronchial/gastric, mucin 5AC, oligomeric mucus/gel-forming, mucin 5AC, oligomeric mucus/gel-forming pseudogene, mucin-5 subtype AC, tracheobronchial, mucin-5AC, TBM, tracheobronchial mucin
Host Species Mouse
Immunogen M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Extracellular Matrix, Infections (Virus Bacteria and Parasites), Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4586
Target Species Human, Mouse, Rat, Bovine, Chicken, Feline, Monkey, Rabbit, Porcine (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.