missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC4, Mouse anti-Human, Clone: MUC4/3105, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79732-20UG
This item is not returnable.
View return policy
Description
MUC4 Monoclonal specifically detects MUC4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| MUC4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Q99102, Q99102 | |
| MUC4 | |
| Recombinant fragment (around aa 1730-1864) of human MUC4 protein (exact sequence is proprietary) | |
| 20 μg | |
| Cancer | |
| 4585 | |
| Store at 4°C. | |
| IgG2b κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| MUC4/3105 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Ascites sialoglycoprotein, ASGP, HSA276359, MUC-4, mucin 4, cell surface associated, mucin 4, tracheobronchial, mucin-4, Pancreatic adenocarcinoma mucin, Testis mucin, Tracheobronchial mucin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction