missing translation for 'onlineSavingsMsg'
Learn More

MUC4, Mouse anti-Human, Clone: MUC4/3084, Novus Biologicals™

Mouse Monoclonal Antibody

£203.00 - £377.00

Specifications

Antigen MUC4
Clone MUC4/3084
Concentration 0.2 mg/mL
Dilution Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18275306
View Documents
Novus Biologicals
NBP2-79731-20UG
20 μg
£203.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18206768
View Documents
Novus Biologicals
NBP2-79731-100UG
100 μg
£377.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MUC4 Monoclonal specifically detects MUC4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

MUC4
0.2 mg/mL
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Unconjugated
Mouse
Cancer
10 mM PBS with 0.05% BSA
Ascites sialoglycoprotein, ASGP, HSA276359, MUC-4, mucin 4, cell surface associated, mucin 4, tracheobronchial, mucin-4, Pancreatic adenocarcinoma mucin, Testis mucin, Tracheobronchial mucin
MUC4
IgG1 κ
Protein A or G purified
MUC4/3084
Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Monoclonal
Purified
RUO
Human
Q99102, Q99102
4585
Recombinant fragment (around aa 1730-1864) of human MUC4 protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.