missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MUC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100199
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MUC3A.
MUC3 is an intestinal glycoprotein distributed in the colon and rectum, and to a lesser extent in breast, lung and salivary tissues. It is involved in conferring mucous visoelasticity (RGD).
Specifications
| MUC3 | |
| Polyclonal | |
| Unconjugated | |
| MUC3A | |
| A630081J09Rik; intestinal mucin; Intestinal mucin-3A; MUC3; MUC3A; MUC-3A; mucin 3; mucin 3, intestinal; mucin 3A; mucin 3A, cell surface associated; Mucin-3; mucin-3A; RIKEN cDNA A630081J09 gene | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4584, 619309 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q02505 | |
| MUC3A | |
| A synthesized peptide derived from human MUC3A(Accession Q02505), corresponding to amino acid residues T1896-T1946. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction