Learn More
MUC3 Mouse anti-Human, Clone: SPM200, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of human MUC3.
Brand: Abnova MAB13115.100ug
Description
The mucin genes encode epithelial glycoproteins, some of which are secreted and some membrane bound. Each of the genes contains at least one large domain of tandemly repeated sequence that encodes the peptide sequence rich in serine and/or threonine residues, which carries most of the O-linked glycosylation (Gendler and Spicer, 1995 [PubMed 7778880]).[supplied by OMIM]
Specifications
| MUC3 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| MUC3/mucin | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 4584/57876 | |
| Store at 4°C. | |
| IgG2a κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| SPM200 | |
| Mouse monoclonal antibody raised against synthetic peptide of human MUC3. | |
| In 10mM PBS (0.05% BSA, 0.05% sodium azide). | |
| MUC3A | |
| A synthetic peptide (conjugated with KLH) corresponding to a 35 amino acid fragment, SIB35, which contains two tandem repeats of human MUC3. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.