Learn More
MUC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27675-1-AP-150UL
 View more versions of this product
Product Code. 16875765
  • £349.00 / 150µL
Description
Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.
Specifications
Specifications
MUC2
Polyclonal
Unconjugated
MUC2
Intestinal mucin 2, MLP, MUC 2, MUC2, Mucin 2, SMUC
Rabbit
Antigen Affinity Chromatography
RUO
17831, 4583
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q02817, Q80Z19
Muc2
MUC2 Fusion Protein Ag25800
150 μL
Primary
Human, Mouse