missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MUC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121759
This item is not returnable.
View return policy
Description
MUC2 Polyclonal Antibody for Western Blot, IHC (P)
Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.
Specifications
| MUC2 | |
| Polyclonal | |
| Unconjugated | |
| MUC2 | |
| 2010015E03Rik; colonic mucin; HH-Muc; intestinal mucin-2; MCM; MLP; muc2; MUC-2; mucin 2; mucin 2, intestinal/tracheal; mucin 2, oligomeric mucus/gel-forming; mucin-2; mucin-like protein; secreted gel-forming mucin; SMUC; wnn | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100127145, 17831 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q80Z19 | |
| MUC2 | |
| Recombinant protein Mucin 2. | |
| 20 μL | |
| Primary | |
| Mouse, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction