missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC2 Mouse anti-Human, DyLight 650, Clone: CCP58, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-33157C-100UL
This item is not returnable.
View return policy
Description
MUC2 Monoclonal specifically detects MUC2 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
| MUC2 | |
| Monoclonal | |
| DyLight 650 | |
| Intestinal mucin-2, MLP, MUC-2, mucin 2, intestinal/tracheal, mucin 2, oligomeric mucus/gel-forming, mucin-2, mucin-like protein, SMUC | |
| A synthetic peptide of 29 amino acids, KYPTTTPISTTTMVTPTPTPTGTQTPTTT from MUC2 protein, coupled to KLH. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| CCP58 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4583 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction