missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC2 Antibody (rMLP/842), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08239AF532
This item is not returnable.
View return policy
Description
MUC2 Monoclonal antibody specifically detects MUC2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| MUC2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| Intestinal mucin-2, MLP, MUC2, MUC-2, mucin 2, intestinal/tracheal, mucin 2, oligomeric mucus/gel-forming, mucin-2, SMUC | |
| A synthetic peptide of 29 amino acids, KYPTTTPISTTTMVTPTPTPTGTQTPTTT from MUC2 protein, coupled to KLH (Uniprot: Q02817) | |
| 100 μg | |
| Cancer, Cellular Markers, Inflammation, Signal Transduction | |
| 4583 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| rMLP/842 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction