missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MUC17 Antibody (Forerunner patent anti-Muc 17) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28802-1mg
This item is not returnable.
View return policy
Description
MUC17 Monoclonal antibody specifically detects MUC17 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| MUC17 | |
| Monoclonal | |
| Unconjugated | |
| Q685J3 | |
| Human | |
| Protein A purified | |
| RUO | |
| 140453 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Forerunner patent anti-Muc 17 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| membrane mucin MUC17, MUC-3, mucin 17, cell surface associated, secreted mucin MUC17, small intestinal mucin MUC3, small intestinal mucin-3 | |
| MUC17 | |
| 1 mg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction