missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC16/CA125 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20077-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody recognizes the C-term of MUC16.
CA 125 determit is present on a high molecular weight, mucin like glycoprotein of high molecular weight. CA 125 has been found on frozen sections of amnion and derivatives of coelomic and mullerian epithelium, including pleura, pericardium and peritoneum. In adult tissues, epithelial cells of fallopian tube, endometrium and endocervix; pancreas, colon, gall bladder, stomach, kidney, apocrine sweat gland and mammary gland. It is also found in mesothelial cell lining of pleura, pericardium and peritoneum. It is found in ovarian tumors of serous, endometrioid or clear cell types and adenocarcinomas of mullerian type.Specifications
| MUC16/CA125 | |
| Polyclonal | |
| Unconjugated | |
| MUC16 | |
| CA 125, CA125, MUC 16, MUC16, Mucin 16 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 94025 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8WXI7 | |
| MUC16 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction