missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MUC15 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15604135
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15604135 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15604135 Supplier Invitrogen™ Supplier No. PA550199

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total MUC15 protein.

May play a role in the cell adhesion to the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUC15
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MUC15
Gene Accession No. Q8C6Z1, Q8N387
Gene Alias 4732460E09; D730046L02Rik; Muc15; MUC-15; mucin 15; mucin 15, cell surface associated; mucin-15; PAS3; PASIII; UNQ750/PRO1481
Gene Symbols MUC15
Host Species Rabbit
Immunogen Synthesized peptide derived from human mucin 15, cell surface associated protein (aa 180-260).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143662, 269328
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.