missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC13 Antibody (MUC13/13111), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44615-100ug
This item is not returnable.
View return policy
Description
MUC13 Monoclonal antibody specifically detects MUC13 in Human samples. It is validated for Immunohistochemistry
Specifications
| MUC13 | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 56667 | |
| Store at 4⪚C. | |
| IgG1 κ |
| Immunohistochemistry | |
| MUC13/13111 | |
| Unconjugated | |
| Down-regulated in colon cancer 1FLJ20063, DRCC1, MUC-13, mucin 13, cell surface associated, mucin 13, epithelial transmembrane, mucin-13, RECC | |
| Recombinant fragment (around aa100-300) of human MUC13 protein (Uniprot: Q9H3R2). Exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction