missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC1 / EMA / CD227 Mouse, Clone: rMUC1/960, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 4582-MSM16-P0
This item is not returnable.
View return policy
Specifications
| MUC1 / EMA / CD227 | |
| Cytoplasmic and cell surface | |
| rMUC1/960 | |
| Unconjugated | |
| NP_001191226 | |
| Mouse | |
| 265-400kDa | |
| Protein A/G Purified | |
| RUO | |
| 4582 | |
| IgG2b κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| MUC1 | |
| Human milk-fat globule membranes (HMFGM) | |
| MCF-7 or MDA-231 cells. Breast, colon, ovarian, endometrial carcinoma. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction