missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC1 (CD227) Mouse anti-Human, Alexa Fluor™ 647, Clone: HMFG2 (also known as HMFG-2, 3.14.A3 or F314AE11), BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566590
This item is not returnable.
View return policy
Description
MUC1 (CD227) Monoclonal antibody specifically detects MUC1 (CD227) in Human samples. It is validated for Flow Cytometry.
Specifications
| MUC1 (CD227) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry | |
| HMFG2 (also known as HMFG-2, 3.14.A3 or F314AE11) | |
| Alexa Fluor 647 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction