missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC1 Antibody (HMFG2) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
£225.00 - £487.00
Specifications
| Antigen | MUC1 |
|---|---|
| Clone | HMFG2 |
| Dilution | Western Blot, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18257513
|
Novus Biologicals
NBP2-62565-0.025MG |
0.025 mg |
£225.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18691749
|
Novus Biologicals
NBP2-62565 |
0.2 mg |
£487.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MUC1 Monoclonal specifically detects MUC1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| MUC1 | |
| Western Blot, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin | |
| MUC1 | |
| IgG λ | |
| This antibody recognises MUC1, a tumor associated antigen which is expressed in >90% ovarian carcinomas. This antigen is a high molecular weight (M, 80,000-200,000) glycoprotein. |
| HMFG2 | |
| Monoclonal | |
| Purified | |
| PBS | |
| 4582 | |
| High-MW glycoprotein of the human milk fat globule. | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title