missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mu Opioid R/OPRM1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-79878
This item is not returnable.
View return policy
Description
mu Opioid R/OPRM1 Polyclonal specifically detects mu Opioid R/OPRM1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| mu Opioid R/OPRM1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| hMOP, KIAA0403, LMOR, MOP, M-OR-1, MOR-1, MOR1MOR, Mu opiate receptor, Mu opioid receptor, mu-type opioid receptor, opioid receptor, mu 1, OPRM | |
| Rabbit | |
| 44.8 kDa | |
| 1 mg | |
| Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
| 4988 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:500 | |
| P35372 | |
| OPRM1 | |
| Synthetic peptide comprising residues 386-400 of the human, mouse and rat MOR-1 protein. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mu Opiod Receptor | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction