missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mu Opioid R/OPRM1 Antibody, Alexa Fluor™ 488 , Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-79878AF488
This item is not returnable.
View return policy
Description
mu Opioid R/OPRM1 Polyclonal antibody specifically detects mu Opioid R/OPRM1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| mu Opioid R/OPRM1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Alexa Fluor 488 | |
| hMOP, KIAA0403, LMOR, MOP, M-OR-1, MOR-1, MOR1MOR, Mu opiate receptor, Mu opioid receptor, mu-type opioid receptor, opioid receptor, mu 1, OPRM | |
| Synthetic peptide comprising residues 386-400 of the human, mouse and rat MOR-1 protein. | |
| 1 mL | |
| Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
| 4988 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction