missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mu Crystallin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31994
This item is not returnable.
View return policy
Description
mu Crystallin Polyclonal specifically detects mu Crystallin in Mouse samples. It is validated for Western Blot.
Specifications
| mu Crystallin | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| CRYM | |
| Recombinant protein encompassing a sequence within the center region of human CRYM. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.7mg/mL | |
| Western Blot 1:500-1:3000 | |
| crystallin, mu, DFNA40NADP-regulated thyroid-hormone binding protein, mu-crystallin homolog, NADP-regulated thyroid-hormone-binding protein, THBP | |
| Rabbit | |
| 34 kDa | |
| 0.1 mL | |
| Vision | |
| 1428 | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction