missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTX3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | MTX3 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18246775
|
Novus Biologicals
NBP2-57083 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18621178
|
Novus Biologicals
NBP2-57083-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
MTX3 Polyclonal specifically detects MTX3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Tekniske data
| MTX3 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| MTX3 metaxin 3 | |
| MTX3 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 345778 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FRLSLGGISPAGQETVDANLQKLTQLVNKESNLIEKMDDNLRQSPQLPPRKLPTLKLTPAEEENNS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel