missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556778
This item is not returnable.
View return policy
Description
Immunogen sequence: DGLEEVQKAE MKAYMELVNN MLLTAELYLQ WCDEATVGEI THARYGSPYP WPLNHILAYQ KQWEVKRKMK AIG Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 93%.
The protein encoded by this gene is highly similar to the metaxin 2 protein from mouse, which has been shown to interact with the mitochondrial membrane protein metaxin 1. Because of this similarity, it is thought that the encoded protein is peripherally associated with the cytosolic face of the outer mitochondrial membrane, and that it is involved in the import of proteins into the mitochondrion. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 7.
Specifications
| MTX2 | |
| Polyclonal | |
| Unconjugated | |
| MTX2 | |
| 1500012G02Rik; metaxin 2; Metaxin2; metaxin-2; Mitochondrial outer membrane import complex protein 2; MNCb-0780; Mtx2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10651 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75431 | |
| MTX2 | |
| Recombinant protein corresponding to Human MTX2. Recombinant protein control fragment (Product #RP-96073). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction