missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTUS2 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £299.00
Specifications
| Antigen | MTUS2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18301462
|
Bio-Techne
NBP3-15522-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18369586
|
Novus Biologicals
NBP3-15522-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MTUS2 Polyclonal antibody specifically detects MTUS2 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| MTUS2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| Cardiac zipper protein, CAZIPTracking protein of 150 kDa, KIAA0774+TIP of 150 kDa, microtubule associated tumor suppressor candidate 2, Microtubule plus-end tracking protein TIP150, TIP150microtubule-associated tumor suppressor candidate 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 49-348 of human MTUS2 (NP_056048.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS, 50% glycerol, pH7.3 | |
| 23281 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title