missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTUS2 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Description
MTUS2 Polyclonal antibody specifically detects MTUS2 in Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | MTUS2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 50% glycerol, pH7.3 |
| Gene Alias | Cardiac zipper protein, CAZIPTracking protein of 150 kDa, KIAA0774+TIP of 150 kDa, microtubule associated tumor suppressor candidate 2, Microtubule plus-end tracking protein TIP150, TIP150microtubule-associated tumor suppressor candidate 2 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 49-348 of human MTUS2 (NP_056048.1). |
| Purification Method | Affinity purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?