missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MTTP Polyclonal Antibody
GREENER_CHOICE

Product Code. 17167923
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17167923 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17167923 Supplier Invitrogen™ Supplier No. PA5109994

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SKIVRRVLKE MVAHNYDRF.

MTTP encodes the large subunit of the heterodimeric microsomal triglyceride transfer protein. Protein disulfide isomerase (PDI) completes the heterodimeric microsomal triglyceride transfer protein, which has been shown to play a entral role in lipoprotein assembly. Mutations in MTP can cause abetalipoproteinemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTTP
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene MTTP
Gene Accession No. O08601, P55157
Gene Alias 1810043K16Rik; ABL; LOW QUALITY PROTEIN: microsomal triglyceride transfer protein large subunit; MGC149819; MGC149820; microsomal triglyceride transfer protein; microsomal triglyceride transfer protein (large polypeptide, 88kDa); microsomal triglyceride transfer protein B; microsomal triglyceride transfer protein large subunit; microsomal triglyceride transfer protein, large subunit; Mtp; MTTP
Gene Symbols MTTP
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17777, 310900, 4547
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.