missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mtTFA Antibody (18G102B2E11), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-71648G
This item is not returnable.
View return policy
Description
mtTFA Monoclonal specifically detects mtTFA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated, Knockdown Validated.
Specifications
| mtTFA | |
| Monoclonal | |
| DyLight 488 | |
| Q00059 | |
| TFAM | |
| Human mtTFA [Swiss-Prot# Q00059] | |
| 0.1 mL | |
| Cell Cycle and Replication, Transcription Factors and Regulators | |
| 7019 | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay, KnockDown | |
| 18G102B2E11 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated, KnockDown Validated | |
| mitochondrial transcription factor A, MtTF1, mtTFA, TCF-6, TCF6L1, TCF6L2Mitochondrial transcription factor 1, TCF6TCF6L3, Transcription factor 6, Transcription factor 6-like 2, Transcription factor 6-like 2 (mitochondrial transcription factor), transcription factor A, mitochondrial | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction