Learn More
Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 298-93-1 |
---|---|
Molecular Formula | C18H16BrN5S |
Molecular Weight (g/mol) | 414.33 |
MDL Number | MFCD00011964,MFCD00066662 |
InChI Key | AZKSAVLVSZKNRD-UHFFFAOYSA-M |
Synonym | thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i |
PubChem CID | 64965 |
ChEBI | CHEBI:53233 |
SMILES | [Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15234654
|
Thermo Scientific Acros
158992500 |
250 mg | Glass bottle |
N/A
|
N/A | ||||
15214654
|
Thermo Scientific Acros
158990010 |
1 g | Glass bottle |
N/A
|
N/A
|
||||
15224654
|
Thermo Scientific Acros
158990050 |
5 g | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
298-93-1 | |
414.33 | |
AZKSAVLVSZKNRD-UHFFFAOYSA-M | |
64965 | |
[Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1 |
C18H16BrN5S | |
MFCD00011964,MFCD00066662 | |
thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i | |
CHEBI:53233 |
Specifications
298-93-1 | |
100.0 | |
97.5% min. (Argentometry) | |
MFCD00011964,MFCD00066662 | |
thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i | |
AZKSAVLVSZKNRD-UHFFFAOYSA-M | |
2-(3,5-diphenyltetrazol-2-ium-2-yl)-4,5-dimethyl-1,3-thiazole;bromide | |
64965 | |
414.32 | |
Thiazolyl blue tetrazolium bromide, 98% |
97.5 | |
195.0°C | |
C18H16BrN5S | |
27,IV,6045 | |
Solubility in water: 10g/L (20°C). Other solubilities: soluble in 5% methanol, soluble in ethanol | |
[Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1 | |
414.33 | |
CHEBI:53233 | |
98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing genetic defects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF exposed o
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 206-069-5
missing translation for 'rtecsNumber' : XF8060000
missing translation for 'tsca' : TSCA
- Thiazolyl blue tetrazolium bromide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H341-Suspected of causing genetic defects.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only