missing translation for 'onlineSavingsMsg'
Learn More

Melford MTT

Product Code. 30018541
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018541 5 g 5g
30018540 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018541 Supplier Melford Supplier No. M920505.0
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Water soluble salt of Tetrazolium.

Water soluble salt of Tetrazolium. Yields a yellowish solution when prepared in media or salt solutions lacking phenol red. By cleavage of the tetrazolium ring by dehydrogenase enzymes, dissolved MTT is converted into insoluble purple formazan.

Specifications

CAS 298-93-1
Molecular Formula C18H16N5SBr
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.